Order Entry
ContactUsLinkComponent
YEAST PLASMID MINIPREP 2 ZYMOPREP 50PREP
Zymoprep™ Yeast Plasmid Miniprep II, Zymo Research
# de catalogue 76212-310
Fournisseur:  Zymo Research
Numéro CAS:  
YEAST PLASMID MINIPREP 2 ZYMOPREP 50PREP
Zymoprep™ Yeast Plasmid Miniprep II, Zymo Research
# de catalogue 76212-310
Fournisseur:  Zymo Research
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Format:
    Spin column
  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Plasmid
  • Sample type:
    Yeast/Fungi
  • Target molecule:
    Plasmid
  • Description:
    Zymoprep™ Yeast Plasmid Miniprep II
  • Size:
    50 Preps
  • Description:
    Zymoprep™ Yeast Plasmid Miniprep II
  • Yield:
    Typically between 0.01-0.3 ng for most 2 µ based plasmids from 1.5 ml overnight cultures

Spécifications

A propos de cet article

The Zymoprep™ Yeast Plasmid Miniprep II used for plasmid isolation from S. pombe.

  • Simple and Efficient
  • No need for glass beads or phenol

System is ideal for low-copy number and hard to isolate plasmids. Eluted plasmid DNA can be used directly for E. Coli transformation, PCR, and Southern blot analysis.

cerevisiae, C. albicans and S. pombe, and any fungi whose cell walls are susceptible to yeast lytic enzyme lysis.

Customers Who Bought This Also Bought