Dippers, PTFE, Dynalon
66 mm (Body), 250 ml, 600 mm
Catalog # 76210-794
Supplier: Dynalon
Fournisseur #: 307024-0250
Jump to:
Product Details & Documents
Totally inert and non-contaminating PTFE dipper sampler.
Extendable screw-in shafts have a steel core for rigidity and are fully encapsulated in PTFE.
Delivery information: The basic unit consists of a beaker with a single 24" (600 mm) shaft. Replacement shaft is available separately.
Spécifications
- Ø:66 mm (Body)
- Capacité:250 ml
- Handle Length:600 mm
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.



