Biox Microtube Rack
# de catalogue 76208-062
Fournisseur: TWD Scientific
Spécifications
- Color:Assorted
- Description:Microtube Rack
- Material:PP
- No. of Holes:80
Spécifications
A propos de cet article
Microcentrifuge Tube Racks are made of a high-strength polypropylene that is chemically resistant to alcohols and mild organic solvents. The molded 5×16, 80-Well array is designed for easy tube identification.
- Polypropylene
- Easy Mark writing area
- Equipped with side handles
- Assorted colors (red, blue, purple, green and yellow)
Microtube racks are designed for use with 1.5 or 2.0 mL microtubes. Racks feature a molded numeric grid for quick identification purposes.
Customers Who Bought This Also Bought