VWR® L-Shape Cell Spreader, Sterile
Catalogue # 76207-748
Fournisseur: VWR International
Spécifications
- Sterility:Sterile
- Color:Green
- Environmental certification:ISO 14001/Certificate No.: 04923E00243R0M
- Cat. no.:76207-748
- Packaging:Bulk packed; 10/bag
Spécifications
À propos de cet article
Disposable L-shaped cell spreaders are ideal for spreading cell or bacterial cultures evenly.
- L-shape allows application on the entire dish or plate surface
- Smooth spreading surface with slight upward return reduces the risk of damage to samples
- Sterilized by electron beam irradiation
- Non-pyrogenic
Spreaders made of polypropylene and pre-sterilized, eliminating the need for flaming or autoclaving.
Certifications: Manufactured in a class 100000 cleanroom environment; ISO13485:2003 and ISO 9001:2008.
Packaging: Individual wrap and bulk packaging available.
Frequently Bought Together