ContactUsLinkComponent
VWR® L-Shape Cell Spreader, Sterile
Catalogue # 76207-748
Fournisseur:  VWR International
undefined
VWR® L-Shape Cell Spreader, Sterile
Catalogue # 76207-748
Fournisseur:  VWR International
Spécifications
  • Sterility:
    Sterile
  • Color:
    Green
  • Environmental certification:
    ISO 14001/Certificate No.: 04923E00243R0M
  • Cat. no.:
    76207-748
  • Packaging:
    Bulk packed; 10/bag
Spécifications
À propos de cet article
Disposable L-shaped cell spreaders are ideal for spreading cell or bacterial cultures evenly.
  • L-shape allows application on the entire dish or plate surface
  • Smooth spreading surface with slight upward return reduces the risk of damage to samples
  • Sterilized by electron beam irradiation
  • Non-pyrogenic
Spreaders made of polypropylene and pre-sterilized, eliminating the need for flaming or autoclaving.
Certifications: Manufactured in a class 100000 cleanroom environment; ISO13485:2003 and ISO 9001:2008.
Packaging: Individual wrap and bulk packaging available.
Frequently Bought Together