ContactUsLinkComponent
Accugreen*Broad Range Dsdna Quantitation Solution 100 Assay
AccuGreen™ High Sensitivity and Broad Range dsDNA Quantitation Kits, Biotium
Catalog # 76204-712
Supplier:  Biotium
CAS Number:  
AccuGreen™ High Sensitivity and Broad Range dsDNA Quantitation Kits, Biotium
Catalog # 76204-712
Supplier:  Biotium
CAS Number:  
Specifications
  • Description:
    AccuGreen™ Broad Range dsDNA quantitation kit
  • Size:
    100 assays
  • Ex/Em:
    500/530 nm
  • Cat. no.:
    76204-712
Specifications
About this item
The AccuGreen™ High Sensitivity and Broad Range dsDNA quantitation kits are designed for use with handheld fluorometers such as the Qubit® fluorometers from Thermo Fisher Scientific.
  • Designed specifically for Thermo’s Qubit® fluorometer
  • Linear range: 0.1 to 100 ng dsDNA (AccuGreen™ High Sensitivity), 2 to 1000 ng dsDNA (AccuGreen™ Broad Range)
  • Sample concentration range: 5 pg/µl to 100 ng/µl (AccuGreen™ High Sensitivity), 0.1 to 1000 ng/µl (AccuGreen™ Broad Range)
  • Green fluorescence emission
  • Replaces Qubit® dsDNA HS and BR assay kits for cost savings
Fluorescent dsDNA quantitation kits for sensitive and selective detection of purified dsDNA samples on handheld fluorometers such as the Qubit® fluorometer. Kits available with or without DNA standards, for users that wish to use their own standards.
Customers Who Bought This Also Bought