ContactUsLinkComponent
Cell-Repellent Plate 96well
CELLSTAR® Cell-Repellent Surface Microplates and Dishes, Greiner Bio-One
# de catalogue 76199-200
Fournisseur:  Greiner Bio-One
Cell-Repellent Plate 96well
CELLSTAR® Cell-Repellent Surface Microplates and Dishes, Greiner Bio-One
# de catalogue 76199-200
Fournisseur:  Greiner Bio-One
Spécifications
  • Sterility:
    Sterile
  • Color:
    Clear
  • Description:
    Cell-Repellent 96-Well Microplate with Lid
  • Material:
    PS
  • Cat. no.:
    76199-200
  • Bottom Style:
    U-Bottom
Spécifications
A propos de cet article
Greiner Bio-One introduces a new surface chemistry designed to effectively inhibit cell adhesion.
  • Nonpyrogenic
  • Free of RNase, DNase, and human DNA
  • Cell-repellent surface
  • Ideal substrate for native cell culture experiments
Achieved through an innovative chemical polymer modification, Greiner Bio-One’s cell-repellent surface does not degrade or leach under common cell culture conditions, rendering an ideal substrate for native cell culture experiments.
The cell-repellent surface is ideal for applications such as spheroid culture of tumor cells, spheroid culture of embryonic cells, embryoid body formation, and suspension culture of semi-adherent and adherent cell lines. All vessels are free of detectable RNase, DNase, and human DNA. nonpyrogenic and noncytotoxic.
Customers Who Bought This Also Bought