Ward’s® Adjustable Volume Micropipettes
Catalog # 470234-628
Supplier: Avantor
Specifications
- Volume:—
- Description:Personal Pipette Set
- Cat. no.:470234-628
- Increments:—
Specifications
About this item
The Wards® Micropipette line is fully autoclavable, helping to prevent cross contamination. The ergonomic design is ambidextrous, providing all users with an excellent operating experience.
- Fully autoclavable
- Slim, lightweight ergonomic design
- Compatible with most universal tip brands
- Tool included for quick and easy recalibration
- Purchase individually, or as one of our available sets
Personal Pipette Set comes with one of each pipette shown, plus a storage carousel.
Classroom Pipette Set comes with eight of each pipette shown, plus eight storage carousels.
Customers Who Bought This Also Bought