Spécifications
- Description:Anti-A and Anti-B
- No. of Students:50
- Cat. no.:470233-600
Spécifications
A propos de cet article
Liquid antiserums are pooled and ready to use.
- Anti-A and B Serums Identify ABO Blood Group
- Anti-Rho (D) Identifies + or - Rh Factor
- No Refrigeration Necessary
- Pooled
- Ready-To-Use
Anti-A and B serums identify ABO blood groups (A, B, AB, or O), and anti-AB provides a check on type O determinations. Anti-Rho (D) identifies + or - Rh factor.
Refrigeration is not necessary. The antiserums are contained in dropper bottles. They are for educational purposes only.