ContactUsLinkComponent
Osmo1* Single-Sample Micro-Osmometer 20 uL test volume
Osmometer, Advanced Instruments
# de catalogue 76193-112
Fournisseur:  Advanced Instruments
Osmo1* Single-Sample Micro-Osmometer 20 uL test volume
Osmometer, Advanced Instruments
# de catalogue 76193-112
Fournisseur:  Advanced Instruments
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Single-Sample Micro-Osmometer, 20 µL Single Sample
  • Cat. no.:
    76193-112
Spécifications
A propos de cet article
Best in class single-sample micro-osmometer that is easy-to-use offers precise, accurate results.
  • Industry leading accuracy and precision
  • One step direct sampling
  • Easy-to-use, intuitive touchscreen user interface
  • Small 20 µL sample size
  • Secure and efficient data management
  • Built in quality control
High-quality, easy-to-use single-sample micro-osmometer for those laboratories who prefer to directly draw and test small sample volumes and want accurate, precise results and the security and efficiency of data management.
Certifications: CE. ETL. RoHS. IVD.
Ordering information: General supplies (sold separately) include Micro-Sample Test Kit [133800], Printer paper (5 rolls) [FLA835], 20 µL East-Eject™ Sampler [3M0825], Replacement sample probe [133060R], 50 mOsm/kg Calibration Standard [3MA005], 850 mOsm/kg Calibration Standard [3MA085], and 2000 mOsm/kg Calibration Standard [3MA200].
Customers Who Bought This Also Bought