Marvelx* Full Kit 125umx70mm
# de catalogue 76181-078
Fournisseur: Upchurch Scientific
Customers Who Bought This Also Bought
Sodium hydroxide 50% (w/w) in aqueous solution, VWR Chemicals BDH®
Fournisseur: VWR International
$104.28 to $1,182.11
Ethanol denatured ACS (denatured with 5% isopropanol and 5% methanol), VWR Chemicals BDH®
Fournisseur: VWR International
$411.94 to $1,303.38
uTIP™ CleanPak™ Pipette Tip Reload System for Universal Fit Pipettes, Biotix Inc
Fournisseur: Biotix
$81.72 to $326.77
Pentane ≥98.0%, Laboratory Reagent, VWR Chemicals BDH®
Fournisseur: VWR International
$500.19 to $1,790.36