Marvelx* Full Kit 25umx70mm
# de catalogue 76181-008
Fournisseur: Upchurch Scientific
Customers Who Bought This Also Bought
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Fournisseur: MilliporeSigma
635,20 $ to 2 541,50 $
Chloroform ≥99.90% stabilized, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Fournisseur: VWR International
438,39 $ to 1 639,42 $
Glycerine ≥99.7%, Laboratory Reagent, VWR Chemicals BDH®
Fournisseur: VWR International
881,80 $ to 2 807,56 $
Target™ All-Plastic Disposable Luer-Lok Syringes, Thermo Scientific
Fournisseur: Thermo Scientific
260,09 $ to 2 400,95 $
VWR® Multistage Gas Regulators with Stainless Steel Diaphragms
Fournisseur: VWR International
1 142,25 $