Specifications
- Volume:20 - 300 µl
- Packed:10 BLISTER Refills of 96 Racked Tips
- Cat. no.:76180-356
- Tip Style:D300 Diamond BLISTER Refill
Specifications
About this item
PIPETMAN® DIAMOND Tips are designed to fit your PIPETMAN and PIPETMAX® perfectly. They deliver accurate and precise pipetting when used in conjunction with Gilson pipettes.
- Individually wrapped racks of tips that are convenient, clean, and eco-friendly
- Ready to use, just peel the lid and insert into the empty box
- Uses 45% less plastic than regular rack box
- Fully autoclavable, available as sterilized and filter tips
Smart way to fefill:
BLISTER REFILLS contain individually wrapped racks of tips that are convenient, clean, and eco-friendly. The racks are fully autoclavable and are color-coded for easy identification and compliance with Good Laboratory Practice (GLP). BLISTER REFILLS are available as autoclavable, sterilized, or sterilized filter tips. PIPETMAN DIAMOND filter tips contain a high-density polyethylene filter to protect against aerosol-born contamination and do not contain any sealing additives that can be released into your sample so you can be confident in your results.
Accurate and precise:
Using PIPETMAN DIAMOND Tips ensures maximum accuracy and precision and delivers consistent results. The perfect air-tight seal prevents leaks. The tips are made of pure polypropylene to deliver the correct volume and ensure complete dispensing. Volume graduations make it easy to check the amount aspirated visually.
Convenient:
Ready to use; peel the lid and insert it into an empty box.
Gilson tips are manufactured in a cleanroom environment, ensuring they are free of detectable contaminants that can affect your results. From mold to bench, our tips are certified and traceable to meet the most stringent demands in many applications.