ContactUsLinkComponent
CARROUSEL* Pipette Stand, For DISTRIMAN*, Holds up to 7 pipettes
CARROUSEL™ Pipette Stand, Gilson
# de catalogue 76178-564
Fournisseur:  GILSON, INC.
Product available on GSA Advantage®
CARROUSEL* Pipette Stand, For DISTRIMAN*, Holds up to 7 pipettes
CARROUSEL™ Pipette Stand, Gilson
# de catalogue 76178-564
Fournisseur:  GILSON, INC.
Product available on GSA Advantage®
Spécifications
  • Color:
    White
  • Description:
    Carousel Pipettor Stand (Holds 7 Gilson single or multichannel pipettes)
  • Cat. no.:
    76178-564
Spécifications
A propos de cet article
The CARROUSEL™ Pipette Stand holds up to seven Gilson single or multichannel pipettes, protecting them from contamination.
  • Pipettes hang vertically from the rack, preventing liquids from entering and contaminating the pipette shaft
  • Made from chemically resistant polypropylene and polycarbonate
  • Stand’s heavy base provides added stability
The rotating head of the CARROUSEL Stand provides quick access to pipettes while keeping your benchtop decluttered. Its space-saving design frees your bench space for more important tasks at hand.
Customers Who Bought This Also Bought