CARROUSEL™ Pipette Stand, Gilson
# de catalogue 76178-564
Fournisseur: GILSON, INC.
Product available on GSA Advantage®
Spécifications
- Color:White
- Description:Carousel Pipettor Stand (Holds 7 Gilson single or multichannel pipettes)
- Cat. no.:76178-564
Spécifications
A propos de cet article
The CARROUSEL™ Pipette Stand holds up to seven Gilson single or multichannel pipettes, protecting them from contamination.
- Pipettes hang vertically from the rack, preventing liquids from entering and contaminating the pipette shaft
- Made from chemically resistant polypropylene and polycarbonate
- Stand’s heavy base provides added stability
The rotating head of the CARROUSEL Stand provides quick access to pipettes while keeping your benchtop decluttered. Its space-saving design frees your bench space for more important tasks at hand.
Customers Who Bought This Also Bought