Order Entry
ContactUsLinkComponent
Expert* E10 Gel Loading Round Tip 0.1 - 10 uL
EXPERT™ Universal Fit Gel-Loading Pipette Tips, Gilson
# de catalogue 76178-450
Fournisseur:  GILSON, INC.
EXPERT™ Universal Fit Gel-Loading Pipette Tips, Gilson
# de catalogue 76178-450
Fournisseur:  GILSON, INC.

Spécifications

  • Length:
    45.72 mm
  • Volume:
    0.1 - 10 µl
  • Sterile:
    -
  • Tip Style:
    E10, round, graduated (2 µl), ext.Ø 0.17 mm

Spécifications

A propos de cet article

EXPERT™ tips are designed to fit leading pipette brands.

  • Clear packaging allows for easy tip identification
  • Economical and easy-to-use
  • Tips feature graduations at varying volumes, allowing for quick-checks when aspirating samples

Sterilized EXPERT™ tips are free of human DNA, RNase, DNase, PCR inhibitors and pyrogens.

Certifications: Conforms with ISO 9001:2008.
Customers Who Bought This Also Bought