PIPETMAN® L Fixed Volume Single Channel Pipettor, Gilson®
# de catalogue 76178-414
Fournisseur: GILSON, INC.
Product available on GSA Advantage®
Spécifications
- Volume:20 µl
- Description:Model F20L
Spécifications
A propos de cet article
Fixed volume air-displacement pipettor PIPETMAN® L.
PIPETMAN® L fixed volume pipettors complement the PIPETMAN® L single and multichannel models by offering a comfortable, lightweight line of devices where the volume can never accidently be changed. The graduated cover can be readjusted by users in the laboratory for dense and viscous fluids (once reset, the new volume cannot be accidentally changed). The PIPETMAN® L fixed volume series is only available with plastic tip ejectors. For convenient access to the serviceable parts, the tip ejector is equipped with an easy-release clip ejector.
Thirteen robust models cover a large volume range, from 1 to 1000 µl, all with Gilson's legendary accuracy and precision.
Certifications: Gilson maximum permissible errors are guaranteed only when PIPETMAN® pipettors are used with the recommended PIPETMAN® Diamond Tips.
Customers Who Bought This Also Bought