ContactUsLinkComponent
Expert* EF200ST Sterilized Filter Tip 1 - 200 uL
EXPERT™ Universal Fit Filter Tips, Gilson
Catalog # 76178-376
Supplier:  GILSON, INC.
Expert* EF200ST Sterilized Filter Tip 1 - 200 uL
EXPERT™ Universal Fit Filter Tips, Gilson
Catalog # 76178-376
Supplier:  GILSON, INC.
Specifications
  • Volume:
    1 - 200 µl
  • Length:
    58.57 mm
  • Packed:
    4 Packs/case
  • Cat. no.:
    76178-376
  • Sterile:
    +
  • Tip Style:
    Sterilized Filter Tips, EF200ST
Specifications
About this item
EXPERT Filter Tips feature a patented, low-retention polymer that creates a hydrophobic surface to minimize sample binding when dispensing.
  • Economical and easy-to-use
  • Clear packaging allows you to easily identify the tips
  • Tips feature graduations at varying volumes allowing for quick-checks when aspirating samples
Sterilized EXPERT Tips are free of Human DNA, RNase, DNase, PCR inhibitors and pyrogens.
Certifications: Conforms with ISO 9001:2008.
Customers Who Bought This Also Bought