EXPERT™ Universal Fit Filter Tips, Gilson
# de catalogue 76178-328
Fournisseur: GILSON, INC.
Spécifications
- Length:102.2 mm
- Volume:50 - 1250 µl
- Packed:8 Packs/case
- Filter:+
- Sterile:+
- Tip Style:XL Sterilized Filter Tips, EF1000ST
Spécifications
A propos de cet article
EXPERT Filter Tips feature a patented, low-retention polymer that creates a hydrophobic surface to minimize sample binding when dispensing.
- Economical and easy-to-use
- Clear packaging allows you to easily identify the tips
- Tips feature graduations at varying volumes allowing for quick-checks when aspirating samples
Sterilized EXPERT Tips are free of Human DNA, RNase, DNase, PCR inhibitors and pyrogens.
Certifications: Conforms with ISO 9001:2008.
Customers Who Bought This Also Bought