ContactUsLinkComponent
Expert* EF1000ST XL Sterilized Filter Tip 50 - 1250 uL
EXPERT™ Universal Fit Filter Tips, Gilson
# de catalogue 76178-328
Fournisseur:  GILSON, INC.
EXPERT™ Universal Fit Filter Tips, Gilson
# de catalogue 76178-328
Fournisseur:  GILSON, INC.
Spécifications
  • Length:
    102.2 mm
  • Volume:
    50 - 1250 µl
  • Packed:
    8 Packs/case
  • Filter:
    +
  • Sterile:
    +
  • Tip Style:
    XL Sterilized Filter Tips, EF1000ST
Spécifications
A propos de cet article
EXPERT Filter Tips feature a patented, low-retention polymer that creates a hydrophobic surface to minimize sample binding when dispensing.
  • Economical and easy-to-use
  • Clear packaging allows you to easily identify the tips
  • Tips feature graduations at varying volumes allowing for quick-checks when aspirating samples
Sterilized EXPERT Tips are free of Human DNA, RNase, DNase, PCR inhibitors and pyrogens.
Certifications: Conforms with ISO 9001:2008.
Customers Who Bought This Also Bought