ContactUsLinkComponent
Expert* Pipette tip E10 Tipack 0.1 - 10 uL
PIPETMAN® Expert Tipack™ Racked Tips, Gilson®
Catalog # 76178-312
Supplier:  GILSON, INC.
Product available on GSA Advantage®
Expert* Pipette tip E10 Tipack 0.1 - 10 uL
PIPETMAN® Expert Tipack™ Racked Tips, Gilson®
Catalog # 76178-312
Supplier:  GILSON, INC.
Product available on GSA Advantage®
Specifications
  • Volume:
    0.1 - 10 µl
  • Packed:
    4800 Tips
  • Cat. no.:
    76178-312
  • Tip Style:
    Tipack™ Case E10
Specifications
About this item
One tip, any pipettor. Manufactured in a sterile, cleanroom environment using the purest raw materials and state-of-the-art techniques, users can be assured they will receive a high-quality product offering consistent performance.
  • Ready to use
  • Expert Tips are easy to pick up directly from the rack
  • Tipack™ racks are fully autoclavable and sealed with a security sticker
  • Racks are color-coded to differentiate tip models and indicate the compatible Gilson® pipettor model
To help prevent contamination, the 96 tips are loaded without manual intervention. Tipack™ racks are sealed with a security sticker and are fully autoclavable. They will resist being autoclaved several times. Using PIPETMAN® Tips is the only way to ensure accuracy and reproducibility across the full volume range of PIPETMAN® pipettors.
Packaging: Supplied in convenient and steady Tipack™ racks, come as 10 packs of 96 tips.
Customers Who Bought This Also Bought