PIPETMAN Diamond Tips Empty Racks, Gilson®
# de catalogue 76178-268
Fournisseur: GILSON, INC.
Product available on GSA Advantage®
Spécifications
- Description:Diamond Tips, Rack, Empty, D300 Tips
- Cat. no.:76178-268
- For Use With:20-300 µL PIPETMAN EXPERT Tips
Spécifications
A propos de cet article
These empty racks can be used for ECOPACK and EASYPACK loose tips.
Customers Who Bought This Also Bought