ContactUsLinkComponent
DistriTips* Maxi ST Syringe 12.50 mL
DistriTip® Sterilized Syringe Tips for DISTRIMAN® Repetitive Pipettor, Gilson®
# de catalogue 76178-258
Fournisseur:  GILSON, INC.
Product available on GSA Advantage®
DistriTips* Maxi ST Syringe 12.50 mL
DistriTip® Sterilized Syringe Tips for DISTRIMAN® Repetitive Pipettor, Gilson®
# de catalogue 76178-258
Fournisseur:  GILSON, INC.
Product available on GSA Advantage®
Spécifications
  • Volume:
    10 - 125 µl
  • Tip Style:
    Mini Sterilized DistriTip® Syringe Tips
Spécifications
A propos de cet article
The best quality to guarantee repetitive distribution results.
DistriTip® tips are polypropylene syringes with easy-to-read level indicators. Three models cover the entire volume range from 1 µl to 12.5 ml. More than 100 aliquots can be dispensed from each syringe. These sterilized syringe tips are individually wrapped and certified free of RNase, DNase, DNA, RNA, and protease. Used with the DISTRIMAN® positive-displacement repetitive pipettor, DistriTip® tips provide more accurate and precise measurements for viscous, dense, volatile, or hot liquids.
DistriTip® Syringe Tips are specifically designed for use with the DISTRIMAN® Repetitive Pipettor.
Customers Who Bought This Also Bought