ContactUsLinkComponent
DistriTips* Maxi Syringe 12.50 mL
DistriTip® Syringe Tips for DISTRIMAN® Repetitive Pipettor, Gilson®
# de catalogue 76178-250
Fournisseur:  GILSON, INC.
Product available on GSA Advantage®
DistriTips* Maxi Syringe 12.50 mL
DistriTip® Syringe Tips for DISTRIMAN® Repetitive Pipettor, Gilson®
# de catalogue 76178-250
Fournisseur:  GILSON, INC.
Product available on GSA Advantage®
Spécifications
  • Volume:
    1 - 125 µl
  • Tip Style:
    Micro DistriTip® syringe tips
Spécifications
A propos de cet article
The best quality to guarantee repetitive distribution results
DistriTip® tips are polypropylene syringes with easy-to-read level indicators. Three models cover the entire volume range from 1 µl to 1.25 ml. More than 100 aliquots can be dispensed from each syringe. Used with the DISTRIMAN® positive-displacement repetitive pipettor, DistriTip® tips provide more accurate and precise measurements for viscous, dense, volatile, or hot liquids.
DistriTip® Syringe Tips are specifically designed for use with the DISTRIMAN® Repetitive Pipettor.
Customers Who Bought This Also Bought