ContactUsLinkComponent
PIPETMAN* TIPACK* Diamond Tip 0.1 - 10uL
PIPETMAN® Diamond Tips, Sterilized Filter Tipack™ Racks, Gilson®
Catalog # 76178-212
Supplier:  GILSON, INC.
Product available on GSA Advantage®
undefined
PIPETMAN® Diamond Tips, Sterilized Filter Tipack™ Racks, Gilson®
Catalog # 76178-212
Supplier:  GILSON, INC.
Product available on GSA Advantage®
Specifications
  • Volume:
    0.1 - 10 µl
  • Packed:
    10 Tipack™ Racks of 96 Tips
  • Cat. no.:
    76178-212
  • Sterile:
    +
  • Tip Style:
    DFL10ST Diamond Filter Tipack™
Specifications
About this item
Highest quality Diamond Tips for superior results.
Tipack™ racks are supplied as 10 packs of 96 tips
Autoclaving conditions: Moist heat/121 °C/20 min/1 bar. Remove blister packaging before autoclaving.
These PIPETMAN® Filter Tips play a critical role in preventing aerosol-borne contamination. High-density polyethylene filter stops particles down to 1 µm, providing an effective barrier against aerosols. PIPETMAN® Filter Tips are designed to meet the most stringent demands in various fields and applications such as PCR, molecular biology, and cell technology. Sterilized filter tips are certified free of detectable RNase, DNase, DNA, RNA, and protease.
Frequently Bought Together