PIPETMAN® Diamond Tips, Autoclavable, in Tipack™ Racks, Gilson®
Catalog # 76178-204
Supplier: GILSON, INC.
Product available on GSA Advantage®
Specifications
- Volume:2 - 200 µl
- Packed:10 Tipack™ Racks of 96 Tips
- Cat. no.:76178-204
- Tip Style:D200ST Sterile Diamond Tipack™
Specifications
About this item
When presented in convenient, steady, Tipack™ racks, Diamond Tips are easy to pick up directly from the rack.
- To help prevent contamination, the 96 tips are loaded without manual intervention
- Autoclaving conditions: Moist heat/121 °C/20 min/1 bar
- Fully autoclavable and sealed with a security sticker
- Color-coded to differentiate tip models
- Indicate the compatible Gilson® pipettor models
Tipack™ racks are sealed with a security sticker, fully autoclavable, and they will resist being autoclaved several times. Using Gilson PIPETMAN® Tips is the only way to ensure accuracy and reproducibility across the full volume range of PIPETMAN® pipettors.
Frequently Bought Together