ContactUsLinkComponent
PIPETMAN* TIPACK* Diamond Tip 2 - 200uL
PIPETMAN® Diamond Tips, Autoclavable, in Tipack™ Racks, Gilson®
Catalog # 76178-204
Supplier:  GILSON, INC.
Product available on GSA Advantage®
undefined
PIPETMAN® Diamond Tips, Autoclavable, in Tipack™ Racks, Gilson®
Catalog # 76178-204
Supplier:  GILSON, INC.
Product available on GSA Advantage®
Specifications
  • Volume:
    2 - 200 µl
  • Packed:
    10 Tipack™ Racks of 96 Tips
  • Cat. no.:
    76178-204
  • Tip Style:
    D200ST Sterile Diamond Tipack™
Specifications
About this item
When presented in convenient, steady, Tipack™ racks, Diamond Tips are easy to pick up directly from the rack.
  • To help prevent contamination, the 96 tips are loaded without manual intervention
  • Autoclaving conditions: Moist heat/121 °C/20 min/1 bar
  • Fully autoclavable and sealed with a security sticker
  • Color-coded to differentiate tip models
  • Indicate the compatible Gilson® pipettor models
Tipack™ racks are sealed with a security sticker, fully autoclavable, and they will resist being autoclaved several times. Using Gilson PIPETMAN® Tips is the only way to ensure accuracy and reproducibility across the full volume range of PIPETMAN® pipettors.
Frequently Bought Together