MICROMAN® Sterilized Capillary Pistons, Gilson®
# de catalogue 76178-156
Fournisseur: GILSON, INC.
Product available on GSA Advantage®
Spécifications
- Packed:192 Pistons
- Cat. no.:76178-156
- Tip Style:Sterilized Capillary Pistons CP100ST
- For Use With:MICROMAN® M100E
Spécifications
A propos de cet article
Capillary Pistons are designed, developed, and manufactured to fit the MICROMAN® perfectly.
Capillary pistons (CPs) provide 100% protection against contamination. With the disposable piston located inside the capillary tip, users can be assured that no harmful samples will contaminate the pipettor. The capillaries are made of pure polypropylene, are free of dyes, and fit even in the narrowest and deepest tubes.
They ensure optimal performance of the MICROMAN® pipettor across the full volume range.
Certifications: ISO 8655-2.
Customers Who Bought This Also Bought