ContactUsLinkComponent
PIPETMAN* EASYPACK Tip 2 - 200uL
PIPETMAN® Diamond Tips EasyPack™, Gilson®
# de catalogue 76178-152
Fournisseur:  GILSON, INC.
Product available on GSA Advantage®
PIPETMAN* EASYPACK Tip 2 - 200uL
PIPETMAN® Diamond Tips EasyPack™, Gilson®
# de catalogue 76178-152
Fournisseur:  GILSON, INC.
Product available on GSA Advantage®
Spécifications
  • Volume:
    2 - 200 µl
  • Packed:
    1000 Tips
  • Cat. no.:
    76178-152
  • Tip Style:
    D200 Diamond EasyPack™
Spécifications
A propos de cet article
Highest quality Diamond Tips for superior results.
  • Autoclaving conditions: Moist heat/121 °C/20 min/1 bar
  • EasyPack™ offers an easy way to order PIPETMAN® tips
Each box contains 1000 tips (5 bags of 200 tips/bag, or 1 bag of 1000 tips). Exceptions are the D10 ml tips, which contain 200 tips (4 bags of 50 tips/bag) and the D5000 tips (3 bags of 334 tips).
Bulk packs are sealed with a tamper-proof strip which can easily be opened for quick access to the tips. Bulk packs are closed using the rapid-sealing strip to prevent contamination and loss of tips.
Customers Who Bought This Also Bought