ContactUsLinkComponent
TRIO* Pipette Stand, For PIPETMAN* Classic, Holds up to 3 pipettes
TRIO™ Pipette Stand, Gilson
# de catalogue 76178-108
Fournisseur:  GILSON, INC.
Product available on GSA Advantage®
TRIO* Pipette Stand, For PIPETMAN* Classic, Holds up to 3 pipettes
TRIO™ Pipette Stand, Gilson
# de catalogue 76178-108
Fournisseur:  GILSON, INC.
Product available on GSA Advantage®
Spécifications
  • Color:
    Transparent
  • Description:
    TRIO™ Pipettor Stand (Holds 3)
  • Cat. no.:
    76178-108
Spécifications
A propos de cet article
The rugged TRIO™ Pipette Stand provides a space-saving way to store and protect up to three single and multichannel PIPETMAN® pipettes.
  • Protects up to three valuable single channel pipettes from benchtop contamination
  • Pipettes hang vertically from the rack, preventing liquids from entering and contaminating the pipette shaft
  • Transparent stand lets you easily see other items behind it
Because it is manufactured from chemically resistant materials, the stand can be used in most lab environments.
Customers Who Bought This Also Bought