ContactUsLinkComponent
PIPETMAN* G P1000G Pipette 100-1000ul
PIPETMAN® Single Channel Pipettor, Gilson®
# de catalogue 76178-040
Fournisseur:  GILSON, INC.
Product available on GSA Advantage®
PIPETMAN* G P1000G Pipette 100-1000ul
PIPETMAN® Single Channel Pipettor, Gilson®
# de catalogue 76178-040
Fournisseur:  GILSON, INC.
Product available on GSA Advantage®
Spécifications
  • Volume:
    100 - 1000 µl
  • Description:
    PIPETMAN® P1000, stainless steel tip ejector
  • For Use With:
    D1000, D1200, DF1000 and DF1200 diamond tips
Spécifications
A propos de cet article
Fully adjustable air-displacement pipettor PIPETMAN®.
  • Extremely low pipetting forces
  • Ergonomic and easy to use with premium comfort and performance
PIPETMAN® brings a new level of comfort into Gilson range with a redesigned piston mechanism that reduces the force needed to aspirate and dispense liquids. PIPETMAN® maintains the trusted durability, accuracy, and reliability of the original PIPETMAN® pipettes with comfort.
Certifications: Gilson maximum permissible errors are guaranteed only when PIPETMAN® pipettors are used with the recommended PIPETMAN® Diamond Tips.
Customers Who Bought This Also Bought