PIPETMAN® Single Channel Pipettor, Gilson®
# de catalogue 76178-034
Fournisseur: GILSON, INC.
Product available on GSA Advantage®
Spécifications
- Volume:2 - 20 µl
- Description:PIPETMAN® P20, stainless steel tip ejector
- For Use With:D200 and DF30 diamond tips
Spécifications
A propos de cet article
Fully adjustable air-displacement pipettor PIPETMAN®.
- Extremely low pipetting forces
- Ergonomic and easy to use with premium comfort and performance
PIPETMAN® brings a new level of comfort into Gilson range with a redesigned piston mechanism that reduces the force needed to aspirate and dispense liquids. PIPETMAN® maintains the trusted durability, accuracy, and reliability of the original PIPETMAN® pipettes with comfort.
Certifications: Gilson maximum permissible errors are guaranteed only when PIPETMAN® pipettors are used with the recommended PIPETMAN® Diamond Tips.
Customers Who Bought This Also Bought