Specifications
- Description:Vi-CELL XR Quad Pak, includes 4 reagent packs and 8 bags of 120 sample vials
- Cat. no.:76176-664
Specifications
About this item
Consumables from the Vi-CELL manufacturer ensure optimum performance and results from your Vi-CELL.
- Concentration Control to run daily to ensure proper instrument performance
- Focus Control to perform auto-focus routine
- Sample vials designed to properly fit in the Vi-CELL sample carousel
- Reagent Paks designed to fit in the Vi-CELL XR with color-coded tubing for error-free installation
The Vi-CELL Concentration Control should be run daily to ensure proper instrument performance and results.
The Vi-CELL Focus Control should be used with the Focusing Wizard to automatically check and if necessary, adjust the focus; especially after the instrument has been physically moved to ensure optimum results.
The Vi-CELL sample vials, which hold up to 4 mL, should be used exclusively with the Vi-CELL, as they have been designed specifically for the carousel.
The Vi-CELL Quad Paks are the key to the Vi-CELL automated sample preparation for the Trypan Blue cell viability analysis . Each reagent pak is color coded to match the reagent lines to make replacing the pak easy.