Order Entry
ContactUsLinkComponent
Wireless Tube Rack, 20mm 20-Place
Half Size Wireless Nylon Tube Racks, Globe Scientific
# de catalogue 76170-244
Fournisseur:  Globe Scientific
Wireless Tube Rack, 20mm 20-Place
Half Size Wireless Nylon Tube Racks, Globe Scientific
# de catalogue 76170-244
Fournisseur:  Globe Scientific

Spécifications

  • Dimensions:
    128 x 103 x 83 mm (5 x 4 x 3.3")
  • Color:
    Yellow
  • No. of Holes:
    20 (4×5)
  • Cat. no.:
    76170-244

Spécifications

A propos de cet article

These fully autoclavable racks are molded from reinforced nylon for strength and durability.

  • Single piece, three tier grid design for secure and stable racking of tubes
  • Good temperature and chemical resistance to withstand a variety of lab procedures
  • Large, flat endplates for labeling and easy sample identification
  • Dense nylon material will not float in water bath
  • 15 ml conicals fit in 20 mm racks and 50 ml conicals fit in 30 mm racks
  • Seven colorful options to keep samples and benchtop organized

With no coatings to peel or chip, the wireless tube racks are a great alternative to wire coated racks where the formation of rust can be an issue.

Customers Who Bought This Also Bought