Full Size Wireless Nylon Tube Racks, Globe Scientific
Catalog # 76169-940
Supplier: Globe Scientific
Specifications
- No. of holes:40 (4×10)
- Color:Red
- Dimensions:250 x 102 x 83 mm (9.8 x 4 x 3.3")
- Cat. no.:76169-940
Specifications
About this item
These fully autoclavable racks are molded from reinforced nylon for strength and durability.
- Single piece, three tier grid design for secure and stable racking of tubes
- Good temperature and chemical resistance to withstand a variety of lab procedures
- Large, flat endplates for labeling and easy sample identification
- Dense nylon material will not float in water bath
- 15 ml conicals fit in 20 mm racks and 50 ml conicals fit in 30 mm racks
- Seven colorful options to keep samples and benchtop organized
With no coatings to peel or chip, the wireless tube racks are a great alternative to wire coated racks where the formation of rust can be an issue.
Frequently Bought Together