SiliaPrep* SPE cartridge Diethylamine (WAX-2) 500mg
# de catalogue CA75958-894
Fournisseur: SiliCycle
SiliaPrep* SPE cartridge Diethylamine (WAX-2) 500mg
# de catalogue CA75958-894
Fournisseur: SiliCycle
Customers Who Bought This Also Bought
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Fournisseur: MilliporeSigma
635,20 $ to 2 541,50 $
Sodium hydroxide 40% (w/w) in aqueous solution, VWR Chemicals BDH®
Fournisseur: VWR International
146,85 $ to 576,51 $
Alcohol ≥70%, Reagent Grade (denatured with methanol and isopropanol), VWR Chemicals BDH®
Fournisseur: VWR International
131,08 $ to 405,43 $