SiliaPrep* SPE cartridge PropylsulfonicAcid (SCX-2) 2 g
# de catalogue CA75958-512
Fournisseur: SiliCycle
SiliaPrep* SPE cartridge PropylsulfonicAcid (SCX-2) 2 g
# de catalogue CA75958-512
Fournisseur: SiliCycle
Customers Who Bought This Also Bought
Alcohol ≥70%, Reagent Grade (denatured with methanol and isopropanol), VWR Chemicals BDH®
Fournisseur: VWR International
$131.08 to $405.43
Bromothymol blue sodium salt 0.04% (w/v) in aqueous solution, VWR Chemicals BDH®
Fournisseur: VWR International
$99.14 to $1,086.75
Sodium hydroxide 1.0 N in aqueous solution, VWR Chemicals BDH®
Fournisseur: VWR International
$72.65 to $846.50