VWR® Classic Syringe Tips, Sterile
# de catalogue 76163-604
Fournisseur: VWR International
Spécifications
- Volume:1.25 ml
- Cat. no.:76163-604
- Sterile:+
Spécifications
A propos de cet article
Sterile classic syringe tips are individually wrapped, and certified to be pyrogen-, ATP-, Rnase-, and Dnase-free.
- Ideal for viscous, infectious, and volatile liquids
- Compatible with a variety of pipettors
Ripette®, Ripette®pro, Ripette®genX, eLineTM-Dispenser, Minilab 100, Minilab 101, Minilab 201, Distriman®, StepMate, EasyStep, EasyStep electronic, RepeatOne®, Handrop.
These syringe tips are not compatible with: HandyStep®, Repetman®.
They provide reliable and repeatable results for a wide range of liquids, including viscous, infectious, and volatile substances.
Ordering information: The 25 and 50 ml tips require an adapter (needs to be ordered separately).
Customers Who Bought This Also Bought