ContactUsLinkComponent
VWR* Wards Micropipette+ 1 - 5 ml Red
Ward’s® Adjustable Volume Micropipettes
# de catalogue 470231-604
Fournisseur:  VWR International
VWR* Wards Micropipette+ 1 - 5 ml Red
Ward’s® Adjustable Volume Micropipettes
# de catalogue 470231-604
Fournisseur:  VWR International
Spécifications
  • Volume:
    1000 - 5000 µl
  • Description:
    5000 uL Micropipette
  • Increments:
    50.0 µl
Spécifications
A propos de cet article
The Wards® Micropipette line is fully autoclavable, helping to prevent cross contamination. The ergonomic design is ambidextrous, providing all users with an excellent operating experience.
  • Fully autoclavable
  • Slim, lightweight ergonomic design
  • Compatible with most universal tip brands
  • Tool included for quick and easy recalibration
  • Purchase individually, or as one of our available sets
Personal Pipette Set comes with one of each pipette shown, plus a storage carousel.
Classroom Pipette Set comes with eight of each pipette shown, plus eight storage carousels.
Customers Who Bought This Also Bought