Ward’s® Adjustable Volume Micropipettes
# de catalogue 470231-604
Fournisseur: VWR International
Spécifications
- Volume:1000 - 5000 µl
- Description:5000 uL Micropipette
- Increments:50.0 µl
Spécifications
A propos de cet article
The Wards® Micropipette line is fully autoclavable, helping to prevent cross contamination. The ergonomic design is ambidextrous, providing all users with an excellent operating experience.
- Fully autoclavable
- Slim, lightweight ergonomic design
- Compatible with most universal tip brands
- Tool included for quick and easy recalibration
- Purchase individually, or as one of our available sets
Personal Pipette Set comes with one of each pipette shown, plus a storage carousel.
Classroom Pipette Set comes with eight of each pipette shown, plus eight storage carousels.
Customers Who Bought This Also Bought