ContactUsLinkComponent
DNase I Set
DNase I
Catalog # 77001-900
Supplier:  Zymo Research
CAS Number:  
DNase I Set
DNase I
Catalog # 77001-900
Supplier:  Zymo Research
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Pack type:
    1 set
  • Size:
    250 U
  • Enzyme name:
    DNase I
  • Cat. no.:
    77001-900
Specifications
About this item
DNase I (RNase-free) cuts both double-stranded and single-stranded DNA.
  • Lyophilized enzyme
  • Used for selectively degrading DNA in the presence of RNA
DNase is suited for applications such as nick translation, production of random fragments, cleavage of genomic DNA for footprinting, removal of DNA template after in vitro transcription and removal of DNA from RNA samples prior to applications such as RT-PCR. It is compatible with all of our RNA kits featuring in-column DNase digestion.
Delivery: The set supplied with DNA digestion buffer (4 ml).
Packaging: 1 Set
Customers Who Bought This Also Bought