To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Packaging:Rnase A not included
- Size:1000 units
- Enzyme name:Zymolyase
- Cat. no.:77001-890
Spécifications
A propos de cet article
Yeast lytic enzyme used for digestion of yeast and fungal cell walls.
- Specifications: Lyophilized enzyme provided with Zymolyase Storage buffer.
- Source: Arthrobacter luteus
- Activity: β-1,3-glucanase
- Essential Enzyme: β-1,3-glucan laminaripentaohydrolase
- Optimum pH and Temperature: pH 7.5, 35 °C (lysis of viable yeast), pH 6.5, 45 °C (hydrolysis of yeast glucan)
- Unit Definition: One unit (U) of lytic activity is defined as the amount of enzyme that catalyzes a 10% decrease in optical density at 800 nm (OD800) in 30 minutes
- Assay Condition: Yeast (0.8 - 1.0 OD800) in 50 mM potassium phosphate, pH 7.5, 10 mM 2-mercaptoethanol
Digestion of yeast and fungal cell walls is necessary for many experimental procedures including spheroplasting, immunofluorescence, transformation, protein purification, and others. The use of lytic enzymes like Zymolyase are routinely used for digestion. The Zymolyase from Zymo Research is prepared from Arthrobacter luteus and is 100T equivalent. The storage buffer provided with the lyophilized enzyme has been optimized to confer maximal levels of enzymatic activity. R-Zymolyase also contains RNase A.