Spécifications
- Description:Quick-DNA/RNA Miniprep Plus
- Size:50 Preps
- Binding capacity:100 µg
- Yield:100 µg DNA and RNA (binding capacity), ≥50 µl (elution volume)
- Cat. no.:77001-854
- Details:for Cells, any tissues, blood, biological fluids
Spécifications
A propos de cet article
The Quick-DNA/RNA kits provide a quick method for the isolation of high quality genomic DNA and total RNA from cells, tissue, blood, biological fluids, etc. These kits isolate both genomic DNA and a broad range of RNA species without the use of phenol. Small RNAs and microRNAs ≥17 nt are also recovered – no extra steps required!