Order Entry
ContactUsLinkComponent
OneStep* PCR Inhibitor Removal Kit 50preps
OneStep™ PCR Inhibitor Removal Kits, Zymo Research
# de catalogue 77001-830
Fournisseur:  Zymo Research
Numéro CAS:  
OneStep* PCR Inhibitor Removal Kit 50preps
OneStep™ PCR Inhibitor Removal Kits, Zymo Research
# de catalogue 77001-830
Fournisseur:  Zymo Research
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Format:
    Spin column
  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Total RNA
    Total DNA
  • Sample type:
    RNA preps
    DNA preps
  • Target molecule:
    DNA
    RNA
  • Description:
    OneStep™ PCR Inhibitor Removal Kit
  • Size:
    50 preps
  • Description:
    OneStep™ PCR Inhibitor Removal Kit

Spécifications

A propos de cet article

The OneStep™ PCR Inhibitor Removal Kit allows for the efficient removal of contaminants that can inhibit downstream enzymatic reactions from DNA and RNA preparations.

  • Removes PCR inhibitors such as polyphenolics, humic/fulvic acids, tannins, melanin, etc. from nucleic acid solutions to yield high quality DNA or RNA
  • Fast, one-step procedure for cleaning impure samples prior to PCR, sequencing, reverse transcription (RT), etc

About OneStep™ PCR Inhibitor Removal Kit

The OneStep™ PCR Inhibitor Removal Kit contains all the components needed for efficient removal of contaminants that can inhibit downstream enzymatic reactions (e.g. PCR and RT) from DNA and RNA preparations. The column matrices have been specifically designed for the efficient removal of polyphenolic compounds, humic/fulvic acids, tannins, melanin, etc. from the most impure DNA and RNA preparations. Sample clean-up is as simple as applying, spinning, and recovering a sample from the column.

Customers Who Bought This Also Bought