Innovating Science® Dialysis Tubings
# de catalogue 470231-460
Fournisseur: Ward's Science
Spécifications
- Ø:15.9 mm
- Length:152 m
- Width:25 mm
- Description:Dialysis tubing
Spécifications
A propos de cet article
The semi-permeable membrane, created from a natural source of cellulose, allows passage of particles up to 14000 Da.
- Available in multiple lengths
Flat width and open diameter measurement provided in the ordering table.
Customers Who Bought This Also Bought