ContactUsLinkComponent
PCR Tube Strip 0.2mL
Axygen® PCR Strip Tubes, Corning
# de catalogue 76102-482
Fournisseur:  Corning
PCR Tube Strip 0.2mL
Axygen® PCR Strip Tubes, Corning
# de catalogue 76102-482
Fournisseur:  Corning
Spécifications
  • Capacity:
    0.2 ml
  • Color:
    Clear
  • Description:
    8-Strip PCR tubes with individually attached flat caps
  • Cat. no.:
    76102-482