ContactUsLinkComponent
FailSafe* PCR 2X PreMix H 2.5 ml
FailSafe™ PCR Enzyme and 2X Premix Buffers, PCR Optimization Components, Biosearch Technologies
# de catalogue 76081-668
Fournisseur:  Lucigen
Numéro CAS:  
FailSafe* PCR 2X PreMix H  2.5 ml
FailSafe™ PCR Enzyme and 2X Premix Buffers, PCR Optimization Components, Biosearch Technologies
# de catalogue 76081-668
Fournisseur:  Lucigen
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    FailSafe™ PCR 2X PreMix H
  • Cat. no.:
    76081-668
  • Volume:
    2.5 ml
Spécifications
A propos de cet article
Purchase individual FailSafe™ DNA polymerase and/or FailSafe™ 2X PreMix Buffers here
  • Trusted: The same great performance as the FailSafe™ PCR System, but available as individual components
  • Flexible: Purchase any combination of FailSafe™ enzyme and/or 2X PreMixes you desire
This listing is only for reordering of individual FailSafe™ PCR 2X PreMixes or the standalone FailSafe™ DNA polymerase
Before purchasing these components, we strongly recommend first optimizing your PCR reaction conditions using the complete FailSafe™ PCR System in order to know which components to purchase based on their performance.
See the FailSafe™ PCR Systems webpage for more details on this system and PCR components.
Certifications: For research use only. Not for human or diagnostic use.
Ordering information: Each component listed here can be ordered separately and at any quantity. Each 2X Premix includes dNTPs, buffer, and various amounts of MgCl2 and FailSafe™ PCR Enhancer with Betaine
Customers Who Bought This Also Bought