Spécifications
- Description:FailSafe™ PCR 2X PreMix H
- Cat. no.:76081-668
- Volume:2.5 ml
Spécifications
A propos de cet article
Purchase individual FailSafe™ DNA polymerase and/or FailSafe™ 2X PreMix Buffers here
- Trusted: The same great performance as the FailSafe™ PCR System, but available as individual components
- Flexible: Purchase any combination of FailSafe™ enzyme and/or 2X PreMixes you desire
This listing is only for reordering of individual FailSafe™ PCR 2X PreMixes or the standalone FailSafe™ DNA polymerase
Before purchasing these components, we strongly recommend first optimizing your PCR reaction conditions using the complete FailSafe™ PCR System in order to know which components to purchase based on their performance.
See the FailSafe™ PCR Systems webpage for more details on this system and PCR components.