VWR® Upright Freezer Racks for 15 and 50 ml Centrifuge Tube Boxes
Catalog # 76051-792
Supplier: VWR International
Specifications
- Description:VWR® upright freezer rack, 4 deep and 2 high configuration
- Cat. no.:76051-792
- External Dimensions:24⁵/₁₆×10³/₁₆×5⁷/₈"
- Internal Dimensions:6×5¹/₁₆×5¹³/₁₆"
Specifications
About this item
Racks are for standard cardboard 15- and 50-ml centrifuge tubes boxes.
- Constructed of corrosion resistant stain steel
Ordering information: Security lock devices are available for each rack if required.
Customers Who Bought This Also Bought