ContactUsLinkComponent
VWR* Upright Freezer Rack 464 x 259 x 149 mm
VWR® Upright Freezer Racks for 15 and 50 ml Centrifuge Tube Boxes
# de catalogue 76051-790
Fournisseur:  VWR International
VWR* Upright Freezer Rack 464 x 259 x 149 mm
VWR® Upright Freezer Racks for 15 and 50 ml Centrifuge Tube Boxes
# de catalogue 76051-790
Fournisseur:  VWR International
Spécifications
  • Description:
    VWR® upright freezer rack, 3 deep and 2 high configuration
  • Cat. no.:
    76051-790
  • External Dimensions:
    18¹/₄×10³/₁₆×5⁷/₈"
  • Internal Dimensions:
    6×5¹/₁₆×5¹³/₁₆"
Spécifications
A propos de cet article
Racks are for standard cardboard 15- and 50-ml centrifuge tubes boxes.
  • Constructed of corrosion resistant stain steel
Ordering information: Security lock devices are available for each rack if required.
Customers Who Bought This Also Bought