Order Entry
Canada
ContactUsLinkComponent
Quick-DNA™ Plus Kits, Zymo Research
Catalogue # 77001-372
Fournisseur:  Zymo Research
Numéro CAS:  
undefined
Quick-DNA™ Plus Kits, Zymo Research
Catalogue # 77001-372
Fournisseur:  Zymo Research
Supplier Number:  D4074
Numéro CAS:  

Spécifications

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Total DNA
  • Sample type:
    Blood
    Biofluids (serum, plasma, saliva)
    Cultured cells
    Human/animal cells and tissues
  • Target molecule:
    DNA
  • Description:
    Quick-DNA™ Microprep Plus Kit
  • Size:
    50 Preps
  • Sample size:
    ≤1×106 Cells or 5 mg Tissue
  • Description:
    Quick-DNA™ Microprep Plus Kit
  • Sample capacity:
    5 µg
  • Yield:
    The DNA binding capacity of the well is 5 µg. Typically, mammalian tissues yield: 1-3 µg DNA per mg skeletal, heart, lung, and brain tissues and 3-5 µg DNA per mg liver and kidney. Human whole blood will yield 3-7 µg DNA per 100 µl blood sampled.
  • Cat. no.:
    77001-372

Spécifications

À propos de cet article

DNA from any sample (cells, tissues, biological fluids and more)

  • Quick and easy: Simple 20 minute procedure
  • Highest yield: Recover 3× more DNA
  • Ultra-pure: Ready for qPCR, next-generation sequencing, arrays and more

The Quick-DNA™ plus kits are the easiest method for high yield total DNA extraction (e.g., genomic, mitochondrial, viral) from any biological fluid, cell culture, or solid tissue sample. Innovative reagents and zymo-spin technology allow for ultra-pure and concentrated genomic DNA ˃50 kb to be eluted in as little as 10 µl. Zymo-Spin™ columns ensure no buffer retention. Purified DNA is RNA-free bypassing the need for RNase A treatment and ensuring accurate quantification for applications like library preparations. Isolated DNA is suitable for immediate use in sensitive downstream applications including qPCR, DNA-seq, arrays and methylation analysis.

Frequently Bought Together