Easy Purchase Pack Basik* Autosampler vial 2ml
# de catalogue 76047-156
Fournisseur: MicroSolv Technology Corp
Easy Purchase Pack Basik* Autosampler vial 2ml
# de catalogue 76047-156
Fournisseur: MicroSolv Technology Corp
Customers Who Bought This Also Bought
Acetone ≥99.5% (by GC, corrected for water content), OmniSolv®, Supelco®
Fournisseur: MilliporeSigma
768,73 $ to 3 075,66 $
Sodium hydroxide 0.1 N in aqueous solution, VWR Chemicals BDH®
Fournisseur: VWR International
78,90 $ to 874,91 $
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Fournisseur: MilliporeSigma
635,20 $ to 2 541,50 $
Sodium hydroxide 0.01 N in aqueous solution, VWR Chemicals BDH®
Fournisseur: VWR International
82,69 $ to 978,38 $
Reagent alcohol ACS (denatured with methanol and isopropanol), VWR Chemicals BDH®
Fournisseur: VWR International
260,48 $ to 788,04 $