ContactUsLinkComponent
Phusion* High-Fidelity DNA Polymerase 500 units
Phusion High-Fidelity DNA Polymerase, New England Biolabs
# de catalogue CA101641-012
Numéro CAS:  
Phusion* High-Fidelity DNA Polymerase 500 units
Phusion High-Fidelity DNA Polymerase, New England Biolabs
# de catalogue CA101641-012
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Phusion High-Fidelity DNA Polymerase
  • Size:
    500 units
  • Cat. no.:
    CA101641-012
Spécifications
A propos de cet article
High-Fidelity DNA Polymerases are important for applications in which the DNA sequence needs to be correct after amplification.
Manufactured and quality-controlled at New England Biolabs, Thermo Scientific® Phusion High-Fidelity DNA Polymerase offers both high fidelity and robust performance, and thus can be used for all PCR applications. Its unique structure, a novel Pyrococcus-like enzyme fused with a processivity-enhancing domain, increases fidelity and speed. Phusion DNA Polymerase is an ideal choice for cloning and can be used for long or difficult amplicons. With an error rate > 50-fold lower than that of Taq DNA Polymerase and 6-fold lower than that of Pyrococcus furiosus DNA Polymerase, Phusion is one of the most accurate thermostable polymerases available. Phusion DNA Polymerase possesses 5´→ 3´ polymerase activity, 3´→ 5´ exonuclease activity and will generate blunt-ended products. Phusion DNA Polymerase is supplied with standard 5X Phusion HF Buffer, as well as 5X Phusion GC Buffer, which can be used for complex or GC-rich templates.
Customers Who Bought This Also Bought