ContactUsLinkComponent
OneTaq* Hot Start Master Mix 100 reactions
ONETAQ® Hot Start 2X Master Mix with Standard Buffer, New England Biolabs
# de catalogue CA101640-990
Numéro CAS:  
OneTaq* Hot Start Master Mix 100 reactions
ONETAQ® Hot Start 2X Master Mix with Standard Buffer, New England Biolabs
# de catalogue CA101640-990
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    OneTaq Hot Start 2X Master Mix with Standard Buffer
  • Size:
    100 Reactions
  • Cat. no.:
    CA101640-990
Spécifications
A propos de cet article
OneTaq® Hot Start 2X Master Mix with Standard Buffer is an optimized blend of Taq and Deep VentR ™ DNA Polymerases combined with an aptamer-based inhibitor.
This enzyme blend is ideally suited to routine PCR applications from a variety of templates, including pure DNA solutions, bacterial colonies and cDNA products. The 3´→5´ exonuclease activity of Deep Vent DNA Polymerase increases the fidelity and robust amplification of Taq DNA Polymerase. The hot start nature of the enzyme offers convenience with decreased interference from primer dimers and secondary products. The convenient master mix formulation contains dNTPs, MgCl2 and other buffer components and stabilizers, requiring only the addition of primers and DNA template for robust amplification.
Customers Who Bought This Also Bought