Order Entry
ContactUsLinkComponent
DNA Clean and Concentrator* 5, 50 Preps
DNA Clean and Concentrator™ Kits, Zymo Research
# de catalogue 77001-152
Fournisseur:  Zymo Research
Numéro CAS:  
DNA Clean and Concentrator* 5, 50 Preps
DNA Clean and Concentrator™ Kits, Zymo Research
# de catalogue 77001-152
Fournisseur:  Zymo Research
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Format:
    Spin column
  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Amplified/modified DNA
  • Sample type:
    PCR and other enzymatic reactions
  • Target molecule:
    DNA
  • Description:
    DNA Clean & Concentrator™-5 (capped columns)
  • Size:
    50 preps
  • Description:
    DNA Clean & Concentrator™

Spécifications

A propos de cet article

Clean and concentrate up to 5 μg DNA with ≥6 μL elution in as little as 2 minutes with 0 μL wash residue carryover.

  • 2 minute clean-up protocol with small elution volumes
  • Column design allows DNA to be eluted at high concentrations into minimal volumes of water or TE buffer
  • Eluted DNA is well suited for use in PCR, DNA sequencing, DNA ligation, endonuclease digestion, RNA transcription, radiolabeling, arrays, etc

About DNA Clean & Concentrator™-5

The DNA Clean & Concentrator™-5 product provides purification of up to 5 µg DNA from PCR, endonuclease digestions, DNA modification reactions, isotope/fluorescence labeling reactions, etc. This product facilitate the removal of DNA polymerases, modifying enzymes, RNA polymerases, ligases, kinases, nucleases, phosphatases, and restriction endonucleases, as well as free dNTPs and their analogs including radiolabeled and fluorescent derivatives. Eluted DNA is suitable for PCR, arrays, ligation, sequencing, etc.

Customers Who Bought This Also Bought