Order Entry
ContactUsLinkComponent
Zymoclean* Gel DNA Recovery Kit 200 Preps
Zymoclean™ Gel DNA Recovery Kits, Zymo Research
# de catalogue 77001-146
Fournisseur:  Zymo Research
Numéro CAS:  
Zymoclean* Gel DNA Recovery Kit 200 Preps
Zymoclean™ Gel DNA Recovery Kits, Zymo Research
# de catalogue 77001-146
Fournisseur:  Zymo Research
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    DNA gel bands
  • Sample type:
    Gel matrix
  • Target molecule:
    DNA
  • Description:
    Zymoclean™ Gel DNA Recovery Kit (capped spin columns)
  • Size:
    200 preps
  • Description:
    Zymoclean™ Gel DNA Recovery Kit
  • Yield:
    50 bp to 23 kb
  • Cat. no.:
    77001-146

Spécifications

A propos de cet article

The Zymoclean Gel DNA Recovery Kits provide for the rapid purification of high quality DNA from TAE/TBE-bufffered agarose gels.

  • Quick (15 minute) recovery of ultra-pure DNA from agarose gels.
  • Column design permits DNA elution at high concentrations into minimal volumes (≥ 6 µL).
  • Eluted DNA is well suited for use in DNA ligation, sequencing, labeling, PCR, etc.

About Zymoclean™ Gel DNA Recovery Kit

The Zymoclean™ Gel DNA Recovery Kit provides for the rapid purification of high quality DNA from TAE/TBE-buffered agarose gels. The product features Fast-Spin technology to yield high-quality, purified DNA in just minutes. DNA purified using the Zymoclean™ Gel DNA Recovery kit are perfectly suited for use in DNA ligation reactions, sequencing, DNA labeling reactions, PCR, etc.

Customers Who Bought This Also Bought